Lithium ≥99.9% (metals basis), foil, Width: 100 mm (3.9 in), Thickness: 1.5 mm (0.06 in)

Supplier: Thermo Scientific Chemicals

10767-22 10767-22
CAAA10767-22EA 1264 CAD
AA10767-22 CAAA10767-22
Lithium ≥99.9% (metals basis), foil, Width: 100 mm (3.9 in), Thickness: 1.5 mm (0.06 in)
Lithium
Formula: Li
MW: 6.94 g/mol
Boiling Pt: 1317 °C (1013 hPa)
Melting Pt: 179 °C
Density: 0.53 g/cm³ (20 °C)
MDL Number: MFCD00134051
CAS Number: 7439-93-2
UN: 1415
ADR: 4.3,I
Merck Index: 13,05542

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR