Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 1.5 mm (0.059 in)

Supplier: Thermo Scientific Chemicals

Synonyms: Aluminum

10765-G1 10765-G5
CAAA10765-G1EA 76.63 CAD
CAAA10765-G1 CAAA10765-G5
Aluminium ≥99.999% (metals basis), wire, Puratronic®, annealed, Ø 1.5 mm (0.059 in)
Aluminium

MDL: MFCD00134029


Purity: 99.999% CAS No.: 7429-90-5 Physical Form: ≈4.8g/m Odor: Odorless MDL No.: MFCD00134029

Formula: Al
MW: 26.98 g/mol
Boiling Pt: 2327
Melting Pt: 660
Density: 2.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134029
CAS Number: 7429-90-5
EINECS: 231-072-3
Merck Index: 13,00321

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR