Tellurium 99,9999 Metals basis, lumps typ. 12 mm (0.47 in)

Supplier: Thermo Scientific Chemicals
Danger

10758-18 10758-30
CAAA10758-09EA 166.14 CAD
CAAA10758-09 CAAA10758-18 CAAA10758-30
Tellurium 99,9999 Metals basis, lumps typ. 12 mm (0.47 in)
Tellurium

MDL: MFCD00134062


Purity: 99.9999% CAS No.: 13494-80-9 Odor: Odorless MDL No.: MFCD00134062

Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990
Melting Pt: 450
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3284
ADR: 6.1 III
Merck Index: 12,09273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR