Sulfur 99,999 Metals basis, pieces, Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

10755-09 10755-18 10755-30
CAAA10755-09EA 67.47 CAD
CAAA10755-09 CAAA10755-18 CAAA10755-30
Sulfur 99,999 Metals basis, pieces, Puratronic®
Sulphur

MDL: MFCD00085316


Purity: 99.999% CAS No.: 7704-34-9 Physical Form: Random Sizes Odor: Weak UN No.: 1350 MDL No.: MFCD00085316

Formula: S
MW: 32.07 g/mol
Boiling Pt: 445
Melting Pt: 113…119 °C
Density: 2.36 g/cm³ (20 °C)
Flash Pt: 168
Storage Temperature: Ambient
MDL Number: MFCD00085316
CAS Number: 7704-34-9
EINECS: 231-722-6
UN: 1350
ADR: 4.1 III
Merck Index: 13,09059

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR