Lithium niobate ≥99.998% (metals basis excluding Ta), Puratronic® (Ta 50 ppm)

Supplier: Thermo Scientific Chemicals

Synonyms: Lithium niobium oxide

10741-14 10741-22 10741-36
CAAA10741-14EA 154.84 CAD
CAAA10741-14 CAAA10741-22 CAAA10741-36
Lithium niobate ≥99.998% (metals basis excluding Ta), Puratronic® (Ta 50 ppm)
Lithium niobate

MDL: MFCD00011080


Purity: 99.998% CAS No.: 12031-63-9 Molecular Formula: LiNbO3 Formula Weight: 147.85 Physical Form: Powder Density: 4.3 Melting Point: ca 1275° MDL No.: MFCD00011080

Formula: LiNbO₃
MW: 147.85
Melting Pt: 1257
Density: 4.3
Storage Temperature: Ambient
MDL Number: MFCD00011080
CAS Number: 12031-63-9
EINECS: 234-755-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR