Lithium metaborate 99,997, Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
10739-14
10739-22
10739-36
CAAA10739-14EA
150.57
CAD
CAAA10739-14
CAAA10739-22
CAAA10739-36
Lithium metaborate 99,997, Puratronic®
Lithium metaborate
MDL: MFCD00011089
Purity: 99.997%
CAS No.: 13453-69-5
Molecular Formula: BLiO2
Formula Weight: 49.75
Physical Form: Powder
Density: 1.397
Appearance: White
Odor: Odorless
Melting Point: 845°
MDL No.: MFCD00011089
Formula:
BLiO₂ MW: 49.75 g/mol Boiling Pt: 1719 Melting Pt: 845 Density: 1.397 Storage Temperature: Ambient |
MDL Number:
MFCD00011089 CAS Number: 13453-69-5 EINECS: 236-631-5 Merck Index: 13,05546 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...