Lithium fluoride ≥99.99% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10736-09
10736-18
10736-30
CAAA10736-09EA
129.88
CAD
CAAA10736-09
CAAA10736-18
CAAA10736-30
Lithium fluoride ≥99.99% (metals basis), Puratronic®
Lithium fluoride
MDL: MFCD00011090
Purity: 99.99%
CAS No.: 7789-24-4
Molecular Formula: FLi
Formula Weight: 25.94
Physical Form: Powder
Density: 2.635
Refractive Index: 1.3915
Appearance: White
Odor: Odorless
Melting Point: 845°
Boiling Point: 1680°
Merck Reference: 14,5531
UN No.: 3288
MDL No.: MFCD00011090
Formula:
FLi Boiling Pt: 1680 °C Melting Pt: 845 °C Density: 2.635 Storage Temperature: Ambient |
MDL Number:
MFCD00011090 CAS Number: 7789-24-4 EINECS: 232-152-0 UN: 3288 ADR: 6.1,III Merck Index: 14,5531 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...