Lithium chloride hydrate ≥99.996% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Lithium chloride xhydrate, Hydrochloric acid lithium salt hydrate

10735-14 10735-22
CAAA10735-14EA 284.4 CAD
CAAA10735-14 CAAA10735-22
Lithium chloride hydrate ≥99.996% (metals basis), Puratronic®
Lithium chloride hydrate

MDL: MFCD00149764


Purity: 99.996% CAS No.: 85144-11-2 Molecular Formula: LiCl.xH2O Formula Weight: 42.39(anhy) Physical Form: Crystalline Aggregates Sensitivity: Hygroscopic Appearance: White Odor: Odorless Melting Point: 98° -H2O Merck Reference: 14,5528 MDL No.: MFCD00149764

Formula: ClLi·xH₂O
MW: 60.41
Melting Pt: >98 °C (-H₂O)
Storage Temperature: Ambient
MDL Number: MFCD00149764
CAS Number: 85144-11-2
EINECS: 231-212-3
Merck Index: 14,5528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR