Lithium bromide hydrate ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Lithium bromide xhydrate
10733-14
10733-22
CAAA10733-14EA
229.1
CAD
CAAA10733-14
CAAA10733-22
Lithium bromide hydrate ≥99.995% (metals basis), Puratronic®
Lithium bromide hydrate
MDL: MFCD00149763
Purity: 99.995%
CAS No.: 85017-82-9
Molecular Formula: LiBr.xH2O
Formula Weight: 86.85(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 162-167°
Merck Reference: 14,5526
MDL No.: MFCD00149763
Formula:
BrLi·xH₂O MW: 86.85 g/mol (anhy) Boiling Pt: 1265 °C (1013 hPa) Melting Pt: 162…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149763 CAS Number: 85017-82-9 EINECS: 231-439-8 Merck Index: 14,5526 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...