Lead(IV) oxide ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lead dioxide
10729-09
10729-18
CAAA10729-09EA
304.94
CAD
CAAA10729-09
CAAA10729-18
Lead(IV) oxide ≥99.995% (metals basis), Puratronic®
Lead (IV) oxide
MDL: MFCD00011165
Purity: 99.995%
CAS No.: 1309-60-0
Molecular Formula: PbO2
Formula Weight: 239.19
Physical Form: Powder
Density: 9.38
Melting Point: 290° dec.
Merck Reference: 14,5407
UN No.: 1872
MDL No.: MFCD00011165
Formula:
O₂Pb MW: 239.2 g/mol Melting Pt: 290 (dec.) Density: 9.38 Storage Temperature: Ambient |
MDL Number:
MFCD00011165 CAS Number: 1309-60-0 EINECS: 215-174-5 UN: 1872 ADR: 5.1 III Merck Index: 14,5407 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...