Lead(II) nitrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10727-18
10727-30
CAAA10727-18EA
297.04
CAD
CAAA10727-18
CAAA10727-30
CAAA10727-A1
Lead(II) nitrate ≥99.999% (metals basis), Puratronic®
Lead (II) nitrate
MDL: MFCD00011153
Purity: 99.999%
CAS No.: 10099-74-8
Molecular Formula: Pb(NO3)2
Formula Weight: 331.20
Physical Form: Powder
Density: 4.530
Melting Point: 470° dec.
Merck Reference: 14,5414
UN No.: 1469
MDL No.: MFCD00011153
Formula:
Pb(NO₃)₂ Melting Pt: 470 °C (dec.) Density: 4.530 Storage Temperature: Ambient |
MDL Number:
MFCD00011153 CAS Number: 10099-74-8 EINECS: 233-245-9 UN: 1469 ADR: 5.1,II Merck Index: 14,5414 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...