Lead(II) fluoride ≥99.997% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10723-14
10723-22
CAAA10723-14EA
203.82
CAD
CAAA10723-14
CAAA10723-22
Lead(II) fluoride ≥99.997% (metals basis), Puratronic®
Lead (II) fluoride
MDL: MFCD00011162
Purity: 99.997%
CAS No.: 7783-46-2
Molecular Formula: PbF2
Formula Weight: 245.20
Physical Form: Powder
Density: 8.445
Melting Point: 855°
Boiling Point: 1290°
Merck Reference: 14,5408
UN No.: 2291
MDL No.: MFCD00011162
Formula:
F₂Pb Boiling Pt: 1290 °C Melting Pt: 855 °C Density: 8.445 Storage Temperature: Ambient |
MDL Number:
MFCD00011162 CAS Number: 7783-46-2 EINECS: 231-998-8 UN: 2291 ADR: 6.1,III Merck Index: 14,5408 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...