Lead(II) chloride ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Lead dichloride
10722-14
10722-22
CAAA10722-14EA
216.46
CAD
CAAA10722-14
CAAA10722-22
Lead(II) chloride ≥99.999% (metals basis), Puratronic®
Lead (II) chloride
MDL: MFCD00011157
Purity: 99.999%
CAS No.: 7758-95-4
Molecular Formula: Cl2Pb
Formula Weight: 278.10
Physical Form: Crystalline
Density: 5.85
Appearance: White
Odor: Odorless
Melting Point: 501°
Boiling Point: 951°
Merck Reference: 14,5404
UN No.: 2291
MDL No.: MFCD00011157
Formula:
Cl₂Pb MW: 278.11 g/mol Boiling Pt: 950 Melting Pt: 496 Density: 5.85 Storage Temperature: Ambient |
MDL Number:
MFCD00011157 CAS Number: 7758-95-4 EINECS: 231-845-5 UN: 2291 ADR: 6.1 III Merck Index: 14,5404 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...