Iron(II) oxalate dihydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Iron (II) ethanedioate dihydrate, Oxalic acid iron (II) salt dihydrate, Ferrous oxalate dihydrate, Iron(II) oxalate hydrate
10718-14
10718-22
CAAA10718-14EA
125.14
CAD
CAAA10718-14
CAAA10718-22
CAAA10718-36
CAAA10718-88
Iron(II) oxalate dihydrate ≥99.999% (metals basis), Puratronic®
Iron (II) oxalate dihydrate
MDL: MFCD00150040
Purity: 99.999%
CAS No.: 6047-25-2
Molecular Formula: FeC2O4.2H2O
Formula Weight: 179.90 (143.87anhy)
Physical Form: Powder
Density: 2.28
Melting Point: 190° dec.
MDL No.: MFCD00150040
Formula:
Fe(C₂O₄)·2H₂O MW: 179.9 g/mol Melting Pt: 190 (dec.) Density: 2.28 Storage Temperature: Ambient |
MDL Number:
MFCD00150040 CAS Number: 6047-25-2 EINECS: 208-217-4 UN: 3288 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...