Iron(III) nitrate nonahydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10715-06
10715-14
CAAA10715-06EA
202.24
CAD
CAAA10715-06
CAAA10715-14
Iron(III) nitrate nonahydrate ≥99.999% (metals basis), Puratronic®
Iron (III) nitrate nonahydrate
MDL: MFCD00149708
Purity: 99.999%
CAS No.: 7782-61-8
Molecular Formula: Fe(NO3)3.xH2O (x~9)
Formula Weight: 241.86(anhy)
Physical Form: Crystalline Aggregates
Density: 1.684
Appearance: Pale purple
Odor: Acidic
Melting Point: 47°
Boiling Point: 125° dec.
Merck Reference: 14,4027
UN No.: 1466
MDL No.: MFCD00149708
Formula:
Fe(NO₃)₃·9H₂O Boiling Pt: 125 °C (dec.) Melting Pt: 47 °C Density: 1.684 Storage Temperature: Ambient |
MDL Number:
MFCD00149708 CAS Number: 7782-61-8 EINECS: 233-899-5 UN: 1466 ADR: 5.1,III Merck Index: 14,4027 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...