Indium(III) sulfate hydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

Synonyms: Diindium trisulphate hydrate, Diindium trisulfate hydrate

10711-09 10711-18
CAAA10711-09EA 186.44 CAD
CAAA10711-09 CAAA10711-18
Indium(III) sulfate hydrate ≥99.999% (metals basis), Puratronic®
Indium(III) sulfate hydrate

MDL: MFCD00149747


Purity: 99.999% CAS No.: 13464-82-9 Molecular Formula: In2(SO4)3.xH2O (x≈6) Formula Weight: 517.83(anhy) Physical Form: Lump/Powder Melting Point: 250° dec. MDL No.: MFCD00149747

Formula: O₁₂In₂S₃·H₂O
MW: 517.83 g/mol (anhy)
Melting Pt: 250 °C (dec.)
Density: 3.44 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149747
CAS Number: 207399-02-8
EINECS: 236-689-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR