Indium (III) oxide ≥99.997% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals

Synonyms: Indium trioxide, Diindium trioxide

10709-09 10709-18
CAAA10709-09EA 158 CAD
CAAA10709-09 CAAA10709-18
Indium (III) oxide ≥99.997% (metals basis), Puratronic®
Indium (III) oxide

MDL: MFCD00011060


Purity: 99.997% CAS No.: 1312-43-2 Molecular Formula: In2O3 Formula Weight: 277.64 Physical Form: Powder Density: 7.18 Melting Point: 850° volat. Merck Reference: 14,4952 MDL No.: MFCD00011060

Formula: In₂O₃
MW: 277.64 g/mol
Melting Pt: 715.05
Density: 7.18
Storage Temperature: Ambient
MDL Number: MFCD00011060
CAS Number: 1312-43-2
EINECS: 215-193-9
Merck Index: 14,4952

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR