Indium(III) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10708-06
10708-14
10708-22
CAAA10708-06EA
91.32
CAD
CAAA10708-06
CAAA10708-14
CAAA10708-22
Indium(III) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Indium (III) nitrate hydrate
MDL: MFCD00149742
Purity: 99.999%
CAS No.: 207398-97-8
Molecular Formula: In(NO3)3.xH2O
Formula Weight: 300.83(anhy)
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Off-white
Melting Point: 100° -H2O
UN No.: 1477
MDL No.: MFCD00149742
Formula:
InN₃O₉·H₂O Melting Pt: 100 °C (-H₂O) Storage Temperature: Ambient |
MDL Number:
MFCD00149742 CAS Number: 207398-97-8 EINECS: 237-393-5 UN: 1477 ADR: 5.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...