Indium(I) chloride ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10707-06
10707-14
CAAA10707-06EA
511.92
CAD
CAAA10707-06
CAAA10707-14
Indium(I) chloride ≥99.999% (metals basis), Puratronic®
Indium (I) chloride
MDL: MFCD00148875
Purity: 99.999%
CAS No.: 13465-10-6
Molecular Formula: InCl
Formula Weight: 150.27
Physical Form: Crystalline Lump
Density: 4.19
Melting Point: ca 225°
Boiling Point: 608°
UN No.: 3260
MDL No.: MFCD00148875
Formula:
InCl Boiling Pt: 608 °C Melting Pt: ∼225 °C Density: 4.19 Storage Temperature: Ambient |
MDL Number:
MFCD00148875 CAS Number: 13465-10-6 EINECS: 236-693-3 UN: 3260 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...