Copper(II) sulfate hydrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cupric sulfate hydrate, Cupric sulphate hydrate

10701-14 10701-22
CAAA10701-14EA 237 CAD
CAAA10701-14 CAAA10701-22
Copper(II) sulfate hydrate ≥99.999% (metals basis), Puratronic®
Copper (II) sulphate hydrate

MDL: MFCD00149682 Notes: m.p. 30°C -2H2O, 110°C -4H2O, 250°C anhydrous; ñ 2.286 Soluble in water. Insoluble in alcohol


Purity: 99.999% CAS No.: 23254-43-5 Molecular Formula: CuSO4.xH2O Formula Weight: 159.60(anhy) Physical Form: Crystalline Aggregates Odor: Odorless pH: 4 Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,2653 UN No.: 3077 MDL No.: MFCD00149682

Formula: CuO₄S·H₂O
MW: 177.62
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149682
CAS Number: 23254-43-5
EINECS: 231-847-6
UN: 3077
ADR: 9 III
REACH: 01-2119520566-40
Merck Index: 14,2653

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR