Copper(II) oxide ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Cupric oxide, Copper oxide, Copper monoxide granulated
10700-14
10700-22
10700-36
CAAA10700-14EA
178.54
CAD
CAAA10700-14
CAAA10700-22
CAAA10700-36
Copper(II) oxide ≥99.995% (metals basis), Puratronic®
Copper (II) oxide
MDL: MFCD00010979
Purity: 99.995%
CAS No.: 1317-38-0
Molecular Formula: CuO
Formula Weight: 79.55
Physical Form: Powder
Density: 6.3
Refractive Index: 2.63
Appearance: Black
Odor: Odorless
Melting Point: 1326°
Merck Reference: 14,2646
UN No.: 3077
MDL No.: MFCD00010979
Formula:
CuO MW: 79.55 g/mol Boiling Pt: 2000 Melting Pt: 1326 Density: 6.3 Storage Temperature: Ambient |
MDL Number:
MFCD00010979 CAS Number: 1317-38-0 EINECS: 215-269-1 UN: 3077 ADR: 9 III Merck Index: 14,2646 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...