Copper(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10699-06
10699-14
10699-22
10699-36
CAAA10699-06EA
53.4
CAD
CAAA10699-06
CAAA10699-14
CAAA10699-22
CAAA10699-36
Copper(II) nitrate hydrate ≥99.999% (metals basis), Puratronic®
Copper (II) nitrate hydrate
MDL: MFCD00149669
Fieser: 1,163 2,87 12,231 15,101 18,115 19,110 20,121 21,148
Purity: 99.999%
CAS No.: 13778-31-9
Molecular Formula: CuN2O6.xH2O
Formula Weight: 187.55(anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline Aggregates
Sensitivity: Hygroscopic
Appearance: Blue
Odor: Odorless
Merck Reference: 14,2643
UN No.: 3085
MDL No.: MFCD00149669
Formula:
CuN₂O₆·H₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD00149669 CAS Number: 13778-31-9 EINECS: 221-838-5 UN: 3085 ADR: 5.1,III Merck Index: 14,2643 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...