Cobalt(II,III) oxide ≥99.9985% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tricobalt tetraoxide, Cobaltic tetraoxide
10695-09
10695-18
CAAA10695-09EA
221.2
CAD
CAAA10695-09
CAAA10695-18
Cobalt(II,III) oxide ≥99.9985% (metals basis), Puratronic®
Cobalt (II,III) oxide
MDL: MFCD00010939
Purity: 99.9985%
CAS No.: 1308-06-1
Molecular Formula: Co3O4
Formula Weight: 240.80
Physical Form: Powder
Density: 6.11
Appearance: Black
Melting Point: 900° dec.
Merck Reference: 14,2429
UN No.: 3077
MDL No.: MFCD00010939
Formula:
Co₃O₄ Melting Pt: 900 °C (dec.) Density: 6.11 Storage Temperature: Ambient |
MDL Number:
MFCD00010939 CAS Number: 1308-06-1 EINECS: 215-157-2 UN: 3077 ADR: 9,III Merck Index: 14,2429 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...