Cobalt(II) nitrate hexahydrate ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cobalt dinitrate hexahydrate, Cobaltous nitrate hexahydrate
10694-09
10694-18
CAAA10694-09EA
178.54
CAD
CAAA10694-09
CAAA10694-18
Cobalt(II) nitrate hexahydrate ≥99.999% (metals basis), Puratronic®
Cobalt (II) nitrate hexahydrate
MDL: MFCD00149647
Notes: greater than 74°C dec. to oxide
Purity: 99.999%
CAS No.: 10026-22-9
Molecular Formula: Co(NO3)2.6H2O
Formula Weight: 291.04 (182.94anhy)
Physical Form: Wet Crystalline Aggregates
Density: 1.87
Refractive Index: 1.52
Appearance: Red
Odor: Odorless
Melting Point: 55-56°
Merck Reference: 14,2444
UN No.: 1477
MDL No.: MFCD00149647
Formula:
Co(NO₃)₂·6H₂O MW: 291.03 g/mol Melting Pt: 57 Density: 1.87 Storage Temperature: Ambient |
MDL Number:
MFCD00149647 CAS Number: 10026-22-9 EINECS: 233-402-1 UN: 1477 ADR: 5.1 III Merck Index: 14,2444 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...