Cobalt(II) chloride hexahydrate ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

10692-09 10692-18
CAAA10692-09EA 184.86 CAD
CAAA10692-09 CAAA10692-18
Cobalt(II) chloride hexahydrate ≥99.998% (metals basis), Puratronic®
Cobalt (II) chloride hexahydrate

MDL: MFCD00149652 Notes: 52-56°C -4H2O, 100°C -5H2O, 120-140°C -6H2O Fieser: 1,155 6,127 10,101 14,99 15,97 18,107 19,104 20,115 21,142


Purity: 99.998% CAS No.: 7791-13-1 Molecular Formula: ClCo2.6H2O Formula Weight: 237.93 (129.84anhy) Physical Form: Crystalline Aggregates Density: 1.924 Appearance: Red-purple Odor: Odorless pH: 4.9 Melting Point: 87° Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00149652

Formula: CoCl₂·6H₂O
MW: 237.93 g/mol
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: 87 °C
Density: 1.924
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 14,2437

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR