Chromium(III) oxide ≥99.97% (metals basis), powder, Puratronic® -22 Mesh

Supplier: Thermo Scientific Chemicals
Warning

10688-06 10688-14 10688-22 10688-36
CAAA10688-06EA 90.53 CAD
CAAA10688-06 CAAA10688-14 CAAA10688-22 CAAA10688-36
Chromium(III) oxide ≥99.97% (metals basis), powder, Puratronic® -22 Mesh
Chromium (III) oxide

MDL: MFCD00010949


Purity: 99.97% CAS No.: 1308-38-9 Molecular Formula: Cr2O3 Formula Weight: 151.99 Physical Form: -22 Mesh Powder Density: 5.22 Refractive Index: 2.551 Appearance: Pale to dark green Odor: Odorless Melting Point: 2435° Boiling Point: 3000° Merck Reference: 14,2234 MDL No.: MFCD00010949

Formula: Cr₂O₃
Boiling Pt: 3000 °C
Melting Pt: 2435 °C
Density: 5.22
Storage Temperature: Ambient
MDL Number: MFCD00010949
CAS Number: 1308-38-9
EINECS: 215-160-9
Merck Index: 14,2234

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR