Calcium oxide ≥99.998% (metals basis) (excluding other alkaline earth and alkali metals 130 ppm max), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Quicklime, Calcium oxide from marble, caustic, Lime
10684-04
10684-09
10684-18
10684-30
CAAA10684-04EA
79
CAD
CAAA10684-04
CAAA10684-09
CAAA10684-18
CAAA10684-30
Calcium oxide ≥99.998% (metals basis) (excluding other alkaline earth and alkali metals 130 ppm max), Puratronic®
Calcium oxide
MDL: MFCD00010911
Purity: 99.998%
CAS No.: 1305-78-8
Molecular Formula: CaO
Formula Weight: 56.08
Physical Form: Powder
Density: 3.37
Refractive Index: 1.8300
Sensitivity: Air and Moisture Sensitive
Appearance: White
Odor: Odorless
Melting Point: 2572°
Boiling Point: 2850°
Merck Reference: 14,1686
MDL No.: MFCD00010911
Formula:
CaO MW: 56.08 g/mol Boiling Pt: 2850 °C Melting Pt: 2572 °C Density: 3.37 Storage Temperature: Ambient |
MDL Number:
MFCD00010911 CAS Number: 1305-78-8 EINECS: 215-138-9 UN: 1910 ADR: 8,III Merck Index: 14,1686 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...