Calcium chloride hydrate ≥99.9965% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10680-04 10680-09 10680-18 10680-30
CAAA10680-04EA 84.69 CAD
CAAA10680-04 CAAA10680-09 CAAA10680-18 CAAA10680-30
Calcium chloride hydrate ≥99.9965% (metals basis), Puratronic®
Calcium chloride hydrate

MDL: MFCD00149616 Notes: 30°C -4H2O, 200°C -6H2O


Purity: 99.9965% CAS No.: 7774-34-7 Molecular Formula: CaCl2.xH2O (x≈4-6) Formula Weight: 110.99(anhy) Physical Form: Crystalline Aggregates Density: 1.71 Merck Reference: 14,1659 MDL No.: MFCD00149616

Formula: CaCl₂·6H₂O
Melting Pt: 29.54 °C
Density: 1.71
Storage Temperature: Ambient
MDL Number: MFCD00149616
CAS Number: 7774-34-7
EINECS: 607-129-7
Merck Index: 14,1659

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR