Cadmium(II) sulfate hydrate ≥99.996%, Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10675-09
10675-18
CAAA10675-09EA
122.45
CAD
CAAA10675-09
CAAA10675-18
Cadmium(II) sulfate hydrate ≥99.996%, Puratronic®
Cadmium (II) sulphate hydrate
MDL: MFCD00151655
Purity: 99.996%
CAS No.: 15244-35-6
Molecular Formula: 3CdSO4.xH2O (x~8)
Formula Weight: 208.46(anhy)
Physical Form: Crystalline
Density: 3.08
Refractive Index: 1.565
Appearance: White
Odor: Odorless
Melting Point:
Formula:
CdO₄S·H₂O MW: 226.49 g/mol Melting Pt: >40 °C (-H₂O) Density: 3.08 Storage Temperature: Ambient |
MDL Number:
MFCD00151655 CAS Number: 15244-35-6 EINECS: 233-331-6 UN: 2570 ADR: 6.1 III Merck Index: 14,1627 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...