Nickel ≥99.996% (metals basis), powder, Puratronic® -120 mesh (max. 0.05% C)

Supplier: Thermo Scientific Chemicals
Danger

10674-14 10674-22 10674-36
CAAA10674-14EA 170.64 CAD
CAAA10674-14 CAAA10674-22 CAAA10674-36 CAAA10674-88
Nickel ≥99.996% (metals basis), powder, Puratronic® -120 mesh (max. 0.05% C)
Nickel

Purity: 99.996% CAS No.: 7440-02-0 Odor: Odorless MDL No.: MFCD00011137

Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
Density: 8.9 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011137
CAS Number: 7440-02-0
EINECS: 231-111-4
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR