Ruthenium ≤100 ppm ≥99.98% (metals basis excluding Ca), powder, Premion® -22 mesh

Supplier: Thermo Scientific Chemicals
Warning

10667-06 10667-14
CAAA10667-06EA 940.1 CAD
CAAA10667-06 CAAA10667-14
Ruthenium ≤100 ppm ≥99.98% (metals basis excluding Ca), powder, Premion® -22 mesh
Ruthenium

MDL: MFCD00011207


Purity: 99.98% CAS No.: 7440-18-8 Odor: Odorless UN No.: 3089 MDL No.: MFCD00011207

Formula: Ru
MW: 101.07 g/mol
Boiling Pt: 3900 °C (1013 hPa)
Melting Pt: 2310 °C
Density: 12.45 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,08452

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR