Cadmium oxide ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Cadmium (II) oxide
10664-14
10664-22
10664-36
CAAA10664-14EA
144.1
CAD
CAAA10664-14
CAAA10664-22
CAAA10664-36
Cadmium oxide ≥99.998% (metals basis), Puratronic®
Cadmium oxide
MDL: MFCD00010921
Purity: 99.998%
CAS No.: 1306-19-0
Molecular Formula: CdO
Formula Weight: 128.40
Physical Form: Powder
Density: 8.15
Appearance: Red-brown
Odor: Odorless
Melting Point: 1430°
Merck Reference: 14,1623
UN No.: 2570
MDL No.: MFCD00010921
Formula:
CdO MW: 128.41 g/mol Boiling Pt: 1559 Melting Pt: 1426 Density: 8.15 Storage Temperature: Ambient |
MDL Number:
MFCD00010921 CAS Number: 1306-19-0 EINECS: 215-146-2 UN: 2570 ADR: 6.1 III Merck Index: 14,1623 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...