Cadmium(II) hydroxide ≥99.98% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
10662-04
10662-09
10662-14
CAAA10662-04EA
127.03
CAD
CAAA10662-04
CAAA10662-09
CAAA10662-14
Cadmium(II) hydroxide ≥99.98% (metals basis), Puratronic®
Cadmium (II) hydroxide
MDL: MFCD00015995
Purity: 99.98%
CAS No.: 21041-95-2
Molecular Formula: Cd(OH)2
Formula Weight: 146.41
Physical Form: Powder
Density: 4.79
Melting Point: 300° dec.
Merck Reference: 14,1620
UN No.: 2570
MDL No.: MFCD00015995
Formula:
H₂CdO₂ Melting Pt: 300 °C (dec.) Density: 4.79 Storage Temperature: Ambient |
MDL Number:
MFCD00015995 CAS Number: 21041-95-2 EINECS: 244-168-5 UN: 2570 ADR: 6.1,III Merck Index: 14,1620 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...