Cadmium chloride hydrate ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10661-09
10661-18
CAAA10661-09EA
100.17
CAD
CAAA10661-09
CAAA10661-18
Cadmium chloride hydrate ≥99.998% (metals basis), Puratronic®
Cadmium chloride hydrate
MDL: MFCD00149627
Purity: 99.998%
CAS No.: 654054-66-7
Molecular Formula: CdCl2.xH2O (x≈2.5)
Formula Weight: 183.32(anhy)
Physical Form: Crystalline powder
Refractive Index: 1.6513
Sensitivity: Hygroscopic
Merck Reference: 14,1617
UN No.: 2570
MDL No.: MFCD00149627
Formula:
CdCl₂·H₂O Melting Pt: 565 °C Density: 3.327 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149627 CAS Number: 654054-66-7 EINECS: 233-296-7 UN: 2570 ADR: 6.1,II Merck Index: 14,1617 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...