Cadmium(II) carbonate ≥99.998% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
10660-09
10660-18
CAAA10660-09EA
90.22
CAD
CAAA10660-09
CAAA10660-18
Cadmium(II) carbonate ≥99.998% (metals basis), Puratronic®
Cadmium (II) carbonate
MDL: MFCD00010918
Purity: 99.998%
CAS No.: 513-78-0
Molecular Formula: CdCO3
Formula Weight: 172.42
Physical Form: Powder
Density: 4.26
Melting Point: 500° dec.
Merck Reference: 14,1616
UN No.: 2570
MDL No.: MFCD00010918
Formula:
CCdO₃ Melting Pt: 500 °C (dec.) Density: 4.26 Storage Temperature: Ambient |
MDL Number:
MFCD00010918 CAS Number: 513-78-0 EINECS: 208-168-9 UN: 2570 ADR: 6.1,III Merck Index: 14,1616 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...