Bismuth(III) iodide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10656-04
10656-09
10656-18
CAAA10656-04EA
72.84
CAD
CAAA10656-04
CAAA10656-09
CAAA10656-18
Bismuth(III) iodide ≥99.999% (metals basis), Puratronic®
Bismuth (III) iodide
MDL: MFCD00010894
Purity: 99.999%
CAS No.: 7787-64-6
Molecular Formula: BiI3
Formula Weight: 589.69
Physical Form: Lump/Powder
Density: 5.778
Appearance: Dark grey to black
Odor: Acidic
Melting Point: 408°
Boiling Point: ca 500°
Merck Reference: 14,1269
UN No.: 3260
MDL No.: MFCD00010894
Formula:
BiI₃ Boiling Pt: ∼500 °C Melting Pt: 408 °C Density: 5.778 Storage Temperature: Ambient |
MDL Number:
MFCD00010894 CAS Number: 7787-64-6 EINECS: 232-127-4 UN: 3260 ADR: 8,III Merck Index: 14,1269 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...