Barium titanate ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10652-14 10652-22
CAAA10652-06EA 81.73 CAD
CAAA10652-06 CAAA10652-14 CAAA10652-22
Barium titanate ≥99.99% (metals basis), Puratronic®
Barium titanate

MDL: MFCD00003447


Purity: 99.99% CAS No.: 12047-27-7 Molecular Formula: BaTiO3 Formula Weight: 233.19 Physical Form: Powder Density: 5.85 Appearance: White Odor: Odorless Melting Point: 1620° Merck Reference: 14,1000 MDL No.: MFCD00003447

Formula: BaO₃Ti
Melting Pt: 1620 °C
Density: 5.85
Storage Temperature: Ambient
MDL Number: MFCD00003447
CAS Number: 12047-27-7
EINECS: 234-975-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,1000

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR