Barium nitrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10648-06 10648-14 10648-22 10648-36
CAAA10648-06EA 38.55 CAD
CAAA10648-06 CAAA10648-14 CAAA10648-22 CAAA10648-36
Barium nitrate ≥99.999% (metals basis), Puratronic®
Barium nitrate

MDL: MFCD00003442


Purity: 99.999% CAS No.: 10022-31-8 Molecular Formula: BaN2O6 Formula Weight: 261.35 Physical Form: Crystalline Density: 3.24 Appearance: White Melting Point: 592° dec. Merck Reference: 14,983 UN No.: 1446 MDL No.: MFCD00003442

Formula: Ba(NO₃)₂
MW: 261.34 g/mol
Melting Pt: 592 (dec.)
Density: 3.24
Storage Temperature: Ambient
MDL Number: MFCD00003442
CAS Number: 10022-31-8
EINECS: 233-020-5
UN: 1446
ADR: 5.1 II
Merck Index: 14,983

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR