Barium fluoride ≥99.99% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

10647-14 10647-22 10647-36
CAAA10647-14EA 270.18 CAD
CAAA10647-14 CAAA10647-22 CAAA10647-36
Barium fluoride ≥99.99% (metals basis), Puratronic®
Barium fluoride

MDL: MFCD00003450


Purity: 99.99% CAS No.: 7787-32-8 Molecular Formula: BaF2 Formula Weight: 175.36 Physical Form: Powder Density: 4.83 Refractive Index: 1.4741 Odor: Odorless Melting Point: 1353° Boiling Point: 2260° Merck Reference: 14,974 UN No.: 1564 MDL No.: MFCD00003450

Formula: BaF₂
MW: 175.33 g/mol
Boiling Pt: 2260 °C
Melting Pt: 1353 °C
Density: 4.83
Storage Temperature: Ambient
MDL Number: MFCD00003450
CAS Number: 7787-32-8
EINECS: 232-108-0
UN: 1564
ADR: 6.1,III
Merck Index: 14,974

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR