Barium chloride dihydrate ≥99.997% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
10646-14
10646-22
10646-36
CAAA10646-14EA
135.56
CAD
CAAA10646-14
CAAA10646-22
CAAA10646-36
Barium chloride dihydrate ≥99.997% (metals basis), Puratronic®
Barium chloride dihydrate
MDL: MFCD00149154
Purity: 99.997%
CAS No.: 10326-27-9
Molecular Formula: BaCl2.2H2O
Formula Weight: 244.28 (208.25anhy)
Physical Form: Powder
Density: 3.097
Appearance: White
Odor: Odorless
Melting Point: 113° -2H2O
Merck Reference: 14,971
UN No.: 1564
MDL No.: MFCD00149154
Formula:
BaCl₂·2H₂O Boiling Pt: 1560 °C (1013 hPa) Melting Pt: 113 °C (-2H₂O) Density: 3.097 Storage Temperature: Ambient |
MDL Number:
MFCD00149154 CAS Number: 10326-27-9 EINECS: 600-412-6 UN: 1564 ADR: 6.1,III Merck Index: 14,971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...