Antimony(III) oxide ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Antimony trioxide, Diantimony trioxide, di-Antimony trioxide

10641-06 10641-14 10641-22
CAAA10641-06EA 74.42 CAD
CAAA10641-06 CAAA10641-14 CAAA10641-22
Antimony(III) oxide ≥99.999% (metals basis), Puratronic®
Antimony (III) oxide

MDL: MFCD00011214


Purity: 99.999% CAS No.: 1309-64-4 Molecular Formula: Sb2O3 Formula Weight: 291.50 Physical Form: Powder Density: 5.67 Appearance: White Odor: Odorless Melting Point: 656° Boiling Point: 1550° subl. Merck Reference: 14,711 MDL No.: MFCD00011214

Formula: O₃Sb₂
MW: 291.52 g/mol
Boiling Pt: 1425 (subl.)
Melting Pt: 654
Density: 5.67
Storage Temperature: Ambient
MDL Number: MFCD00011214
CAS Number: 1309-64-4
EINECS: 215-175-0
UN: 1549
ADR: 6.1 III
Merck Index: 14,711

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR