Ammonium dihydrogen phosphate ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate
10633-14
10633-22
CAAA10633-14EA
233.84
CAD
CAAA10633-14
CAAA10633-22
Ammonium dihydrogen phosphate ≥99.995% (metals basis), Puratronic®
Ammonium dihydrogen phosphate
MDL: MFCD00003396
Purity: 99.995%
CAS No.: 7722-76-1
Molecular Formula: H6NO4P
Formula Weight: 115.03
Physical Form: Crystalline
Density: 1.803
Appearance: White
Odor: Odorless
Melting Point: 190°
Merck Reference: 14,543
MDL No.: MFCD00003396
Formula:
NH₄H₂PO₄ MW: 115.03 g/mol Melting Pt: 190 Density: 1.803 Storage Temperature: Ambient |
MDL Number:
MFCD00003396 CAS Number: 7722-76-1 EINECS: 231-764-5 Merck Index: 14,543 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...