Ammonium chloride ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
|
Warning
|
10632-18
10632-30
10632-A1
CAAA10632-18EA
149.15
CAD
CAAA10632-18
CAAA10632-30
CAAA10632-A1
Ammonium chloride ≥99.999% (metals basis), Puratronic®
Ammonium chloride
MDL: MFCD00011420
Purity: 99.999%
CAS No.: 12125-02-9
Molecular Formula: NH4Cl
Formula Weight: 53.49
Physical Form: Powder
Density: ≈1.53
Refractive Index: 1.642
Sensitivity: Hygroscopic
Odor: Odorless
Melting Point: 340° subl.
Merck Reference: 14,509
MDL No.: MFCD00011420
Formula:
NH₄Cl Melting Pt: 340 °C (subl.) Density: 1.53 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 Merck Index: 14,509 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...