Magnesium ≥99.98% (metals basis), turnings, Puratronic® 1 cm (0.4 in) & down
Supplier: Thermo Scientific Chemicals
|
Danger
|
10630-06
10630-14
10630-22
10630-36
CAAA10630-06EA
38.24
CAD
CAAA10630-06
CAAA10630-14
CAAA10630-22
CAAA10630-36
Magnesium ≥99.98% (metals basis), turnings, Puratronic® 1 cm (0.4 in) & down
Magnesium
MDL: MFCD00085308
Purity: 99.98%
CAS No.: 7439-95-4
Odor: Odorless
UN No.: 1869
MDL No.: MFCD00085308
Formula:
Mg Storage Temperature: Ambient |
MDL Number:
MFCD00085308 CAS Number: 7439-95-4 EINECS: 231-104-6 UN: 1869 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...