Aluminium oxide ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Scientific Chemicals
10627-14
10627-22
10627-36
CAAA10627-14EA
271.76
CAD
CAAA10627-14
CAAA10627-22
CAAA10627-36
Aluminium oxide ≥99.995% (metals basis), Puratronic®
Aluminium oxide
MDL: MFCD00003424
Purity: 99.995%
CAS No.: 1344-28-1
Molecular Formula: Al2O3
Formula Weight: 101.96
Physical Form: Powder
Density: 3.965
Refractive Index: 1.768
Odor: Odorless
Melting Point: 2045°
Boiling Point: 2980°
Merck Reference: 14,356
MDL No.: MFCD00003424
Formula:
Al₂O₃ Boiling Pt: 2980 °C Melting Pt: 2045 °C Density: 3.965 Storage Temperature: Ambient |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 14,356 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...