Aluminium chloride hexahydrate ≥99.9995% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Aluminum chloride hexahydrate

10622-06 10622-18 10622-30
CAAA10622-06EA 103.02 CAD
CAAA10622-06 CAAA10622-18 CAAA10622-30
Aluminium chloride hexahydrate ≥99.9995% (metals basis), Puratronic®
Aluminium chloride hexahydrate

MDL: MFCD00149134


Purity: 99.9995% CAS No.: 7784-13-6 Molecular Formula: AlCl3.6H2O Formula Weight: 241.43 (133.34anhy) Physical Form: Crystalline Aggregates Density: 2.398 Appearance: White Odor: Odorless pH: 2.5 Melting Point: 100° dec. Merck Reference: 14,337 UN No.: 3260 MDL No.: MFCD00149134

Formula: AlCl₃·6H₂O
MW: 241.43 g/mol
Melting Pt: 100 °C (dec.)
Density: 2.398
Storage Temperature: Ambient
MDL Number: MFCD00149134
CAS Number: 7784-13-6
EINECS: 231-208-1
UN: 3260
ADR: 8,III
Merck Index: 14,337

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR