Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)

Supplier: Thermo Scientific Chemicals

10615-09 10615-18 10615-30
CAAA10615-09EA 219.62 CAD
CAAA10615-09 CAAA10615-18 CAAA10615-30
Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Indium

MDL: MFCD00134048


Purity: 99.9999% CAS No.: 7440-74-6 Odor: Odorless MDL No.: MFCD00134048

Formula: In
Boiling Pt: 2000 °C (1013 hPa)
Melting Pt: 156 °C
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR