Indium ≥99.9995% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Supplier: Thermo Scientific Chemicals
10613-09
10613-18
CAAA10613-09EA
181.7
CAD
CAAA10613-09
CAAA10613-18
Indium ≥99.9995% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Indium
MDL: MFCD00134048
Purity: 99.9995%
CAS No.: 7440-74-6
Odor: Odorless
MDL No.: MFCD00134048
Formula:
In Boiling Pt: 2000 °C (1013 hPa) Melting Pt: 156 °C Density: 7.3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...