Tellurium dioxide, tech.

Supplier: Thermo Scientific Chemicals
Warning

10608-18 10608-30
CAAA10608-18EA 273.34 CAD
CAAA10608-18 CAAA10608-30
Tellurium dioxide, tech.
Tellurium dioxide

MDL: MFCD00011263 Practically insoluble in water. Soluble in NaOH solutions and HCl


CAS No.: 7446-07-3 Molecular Formula: TeO2 Formula Weight: 159.60 Physical Form: Powder Density: 5.8 Odor: Odorless Merck Reference: 14,9123 MDL No.: MFCD00011263

Formula: O₂Te
MW: 159.6 g/mol
Boiling Pt: 1245 °C (1013 hPa)
Melting Pt: 733 °C
Density: 5.8
Storage Temperature: Ambient
MDL Number: MFCD00011263
CAS Number: 7446-07-3
EINECS: 231-193-1
Merck Index: 14,9123

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR