Tellurium 99,5 Metals basis, powder -200 mesh

Supplier: Thermo Scientific Chemicals
Danger

10605-18 10605-30
CAAA10605-18EA 141.73 CAD
CAAA10605-18 CAAA10605-30
Tellurium 99,5 Metals basis, powder -200 mesh
Tellurium

MDL: MFCD00134062


Purity: 99.5% CAS No.: 13494-80-9 Appearance: Grey Odor: Odorless UN No.: 3288 MDL No.: MFCD00134062

Formula: Te
MW: 127.6 g/mol
Boiling Pt: 990
Melting Pt: 450
Density: 8.83 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134062
CAS Number: 13494-80-9
EINECS: 236-813-4
UN: 3288
ADR: 6.1 III
Merck Index: 12,09273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR